Dowex® 50WX2 (H) 100-200 mesh
Supplier: Thermo Fisher Scientific
L13944.18 L13944.30 L13944.0B
L13944.18EA 64.2 EUR
L13944.18 L13944.30 L13944.0B
Dowex® 50WX2 (H) 100-200 mesh
Dowex® 50WX2 (H)
CAS No.: 12612-37-2 MDL No.: MFCD00132722
Storage Temperature: Ambient
MDL Number: MFCD00132722
CAS Number: 12612-37-2