Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L16611.06
L16611.14
L16611.06EA
79.1
EUR
L16611.06
L16611.14
Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride 97%
Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoyl fluoride
Purity: 97%
CAS No.: 34761-47-2
Molecular Formula: C15F30O5
Formula Weight: 830.12
Flash Point: None
Refractive Index: 1.3000
Boiling Point: 200-203°
UN No.: UN3265
MDL No.: MFCD00054660
Formula:
C₁₅F₃₀O₅ Boiling Pt: 200…203 °C Density: 1.80 Storage Temperature: Ambient |
MDL Number:
MFCD00054660 CAS Number: 34761-47-2 ADR: 8,II |