2-Chloro-N-methoxy-N-methylacetamide 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 67442-07-3
Molecular Formula: C4H8ClNO2
Formula Weight: 137.57
Storage Temperature: 2°C to 8°C
Flash Point: 104°(219°F)
Melting Point: 39-41°
Boiling Point: 94-95°/9mm
Beilstein Reference: 1924015
MDL No.: MFCD00134232
Formula:
C₄H₈ClNO₂ Boiling Pt: 94…95 °C (9 mmHg) Melting Pt: 39…41 °C Flash Pt: 104 °C (219 °F) |
MDL Number:
MFCD00134232 CAS Number: 67442-07-3 |