Dichloro(p-cymene)ruthenium(II) dimer 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)
L19126.04 L19126.09 L19126.ME
L19126.04EA 124 EUR
L19126.04 L19126.09 L19126.ME
Dichloro(p-cymene)ruthenium(II) dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer
Purity: 98% CAS No.: 52462-29-0 Molecular Formula: C20H28Cl4Ru2 Formula Weight: 612.40 MDL No.: MFCD00064793
Formula: C₂₀H₂₈Cl₄Ru₂
MW: 612.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064793
CAS Number: 52462-29-0
EINECS: 435-530-5