Phenylselenocyanate 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
L20194.03
L20194.06
L20194.03EA
44.7
EUR
L20194.03
L20194.06
Phenylselenocyanate 98%
Phenylselenocyanate
Purity: 98%
CAS No.: 2179-79-5
Molecular Formula: C7H5NSe
Formula Weight: 182.08
Flash Point:
Formula:
C₇H₅NSe Boiling Pt: 116…117 °C (12 mmHg) Density: 1.484 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00216944 CAS Number: 2179-79-5 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...