DL-Camphor (synthetic) USP
Supplier: Spectrum Chemical
Warning
CA185-110LBBL CA185-2.5KG CA185-25KGBL
SPCMCA185-110LBBLEA 6830 EUR
SPCMCA185-110LBBL SPCMCA185-2.5KG SPCMCA185-25KGBL
DL-Camphor (synthetic) USP
DL-Camphor
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula: C₁₀H₁₆O
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ∼180 °C
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III