Corn oil NF
Supplier: Spectrum Chemical
CO136-200LTBL CO136-4LTGL CO136-25ML
SPCMCO136-200LTBLEA 3850 EUR
SPCMCO136-200LTBL SPCMCO136-4LTGL SPCMCO136-25ML
Corn oil NF
Corn oil
Melting Pt: –10 °C
Density: 0.918 g/cm³ (20 °C)
MDL Number: MFCD00130865
CAS Number: 8001-30-7
EINECS: 232-281-2