Creatine (N-amidinosarcosine) monohydrate
Supplier: Spectrum Chemical
Synonyms: Creatine Hydrate, 2-(1-Methylguanidino)acetic acid monohydrate, Creatine monohydrate, 2-(1-Methylguanidino)acetic acid hydrate, N-Amidinosarcosine monydrate
CR105-12KGBL CR105-1KGBL CR105-50KGBL
SPCMCR105-12KGBLEA 3490 EUR
SPCMCR105-12KGBL SPCMCR105-1KGBL SPCMCR105-50KGBL
Creatine (N-amidinosarcosine) monohydrate
Creatine (N-Amidinosarcosine) monohydrate
Formula: C₄H₉N₃O₂·1H₂O
MW: 149.15 g/mol (131.13 anhy)
MDL Number: MFCD00071582
CAS Number: 6020-87-7
EINECS: 200-306-6
Order Now
Specification Test Results
Assay 99% Min.