Diethyl phthalate NF
Supplier: Spectrum Chemical
Synonyms: Phthalic acid diethyl ester
D1097-200LTBL D1097-20LT D1097-4LTGL
SPCMD1097-200LTBLEA 0 EUR
SPCMD1097-200LTBL SPCMD1097-20LT SPCMD1097-4LTGL
Diethyl phthalate NF
Diethyl phthalate
Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula: C₁₂H₁₄O₄
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 156 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00009111
CAS Number: 84-66-2
EINECS: 201-550-6
Merck Index: 12,07527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d0a6c6b14ead75148c0661141b5e8d00e6fc1a9&vid=2461e9f0e230ca44b5372d1233410fbf47a564c7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 98% Min.