L(-)-Proline USP
Supplier: Spectrum Chemical
Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
P1434-1KG P1434-25KGBL P1434-5KG
SPCMP1434-1KGEA 861 EUR
SPCMP1434-1KG SPCMP1434-25KGBL SPCMP1434-5KG
L(-)-Proline USP
L(-)-Proline
Formula: C₅H₉NO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 220…222 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3
EINECS: 205-702-2
Merck Index: 13,07871