Bismuth(III) citrate, powder USP
Supplier: Spectrum Chemical
Warning
Synonyms: Bismuth citrate
B1973-12KGBL B1973-25KGBL
SPCMB1973-12KGBLEA 0 EUR
SPCMB1973-12KGBL SPCMB1973-25KGBL
Bismuth(III) citrate, powder USP
Bismuth (III) citrate
Form: Powder
Formula: C₆H₅BiO₇
MW: 398.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050817
CAS Number: 813-93-4
EINECS: 212-390-1
Order Now
Specification Test Results
Assay 49% Min.