ESD safety ankle boots, lace-up, uvex 2, 6503
Supplier: UVEX
6503.2/35 6503.2/36 6503.2/37 6503.2/38 6503.2/39 6503.2/40 6503.2/41 6503.2/42 6503.2/43 6503.2/44 6503.2/45 6503.2/46 6503.2/47 6503.2/48 6503.2/49 6503.2/50 6503.2/51 6503.2/52 6503.3/35 6503.3/36 6503.3/37 6503.3/38 6503.3/39 6503.3/40 6503.3/41 6503.3/42 6503.3/43 6503.3/44 6503.3/45 6503.3/46 6503.3/47 6503.3/48 6503.3/49 6503.3/50 6503.3/51 6503.3/52 6503.4/50 6503.4/51 6503.4/52 6503.4/44 6503.4/45 6503.4/46 6503.4/47 6503.4/35 6503.4/36 6503.4/37 6503.4/38 6503.4/39 6503.4/40 6503.4/41 6503.4/42 6503.4/43 6503.4/48 6503.4/49 6503.1/45 6503.1/46 6503.1/47 6503.1/48 6503.1/49 6503.1/50 6503.1/51 6503.1/35 6503.1/36 6503.1/37 6503.1/38 6503.1/39 6503.1/40 6503.1/41 6503.1/42 6503.1/43 6503.1/44 6503.1/52
114-6763EA 148 EUR
114-6763 114-6764 114-6765 114-6766 114-6767 114-6768 114-6769 114-6770 114-6771 114-6772 114-6773 114-6774 114-6775 114-6776 114-6777 114-6778 114-6779 114-6780 UVEX6503.3/35 UVEX6503.3/36 UVEX6503.3/37 UVEX6503.3/38 UVEX6503.3/39 UVEX6503.3/40 UVEX6503.3/41 UVEX6503.3/42 UVEX6503.3/43 UVEX6503.3/44 UVEX6503.3/45 UVEX6503.3/46 UVEX6503.3/47 UVEX6503.3/48 UVEX6503.3/49 UVEX6503.3/50 UVEX6503.3/51 UVEX6503.3/52 UVEX6503.4/50 UVEX6503.4/51 UVEX6503.4/52 UVEX6503.4/44 UVEX6503.4/45 UVEX6503.4/46 UVEX6503.4/47 UVEX6503.4/35 UVEX6503.4/36 UVEX6503.4/37 UVEX6503.4/38 UVEX6503.4/39 UVEX6503.4/40 UVEX6503.4/41 UVEX6503.4/42 UVEX6503.4/43 UVEX6503.4/48 UVEX6503.4/49 UVEX6503.1/45 UVEX6503.1/46 UVEX6503.1/47 UVEX6503.1/48 UVEX6503.1/49 UVEX6503.1/50 UVEX6503.1/51 UVEX6503.1/35 UVEX6503.1/36 UVEX6503.1/37 UVEX6503.1/38 UVEX6503.1/39 UVEX6503.1/40 UVEX6503.1/41 UVEX6503.1/42 UVEX6503.1/43 UVEX6503.1/44 UVEX6503.1/52
ESD safety ankle boots, lace-up, uvex 2, 6503
Shoes and Boots
Metal-free, lightweight, flexible and robust safety shoe made from breathable, water repellent leather upper, soft padded tongue and collar, plus a dual-density PUR sole free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Virtually seam-free construction eliminates pressure points
  • Lightweight two-layer sole with excellent shock absorption and outstanding slip resistance
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Injection-moulded PU scuff cap for additional protection
  • Flexible, metal-free, puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
For medium industrial applications including chemical, transport, logistics, energy and mechanical engineering industries.
Colour: Black/orange
Certifications: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, SRC