Hand disinfectant foam, Deb InstantFOAM® Complete
Supplier: Deb-STOKO
DIF12400 DIF610 DIF610EX IFS1LTFNL DIF12400EX IFS250ML IFS1000ML IFS1LTFDE IFS47ML
115-2272EA 18 EUR
115-2272 115-2275 115-2276 115-2290 115-2273 115-2291 115-2138 115-2289 115-2292
Hand disinfectant foam, Deb InstantFOAM® Complete
Skincare Skin Disinfectants Liquid and Foam Skin Disinfectants
Hypoallergenic, alcohol-based foam sanitiser for use in food industry. Compatible for use with latex and nitrile gloves.
  • Use without water to kill 99,999% of common germs
  • Broad spectrum, highly effective bactericidal (EN1500), virucidal (EN14476), yeasticidal (EN1650), mycobactericidal (EN14348) activity
  • Independently tested and proven to be non tainting to food
  • Quickly and easily spreads across the hands leaving no sticky residues
  • Contains moisturisers to prevent skin dryness
  • Colourant-free and perfume-free
Active ingredients: 80 g Ethanol/100 g product
Caution: Use biocides safely. Always read the label and product information before use.