Dyes, DyLight™
Supplier: Thermo Fisher Scientific
46402 46400 46401 46403 46414 46417 46418 46419 46421 46422 46600 46602 46613 46618 46621 46412 46413 46608 46426 62265 62266 62262 62263 62290 62295 62278 62279 62298
PIER46402EA 442 EUR
PIER46402 PIER46400 PIER46401 PIER46403 PIER46414 PIER46417 PIER46418 PIER46419 PIER46421 PIER46422 PIER46600 PIER46602 PIER46613 PIER46618 PIER46621 PIER46412 PIER46413 PIER46608 PIER46426 PIER62265 PIER62266 PIER62262 PIER62263 PIER62290 PIER62295 PIER62278 PIER62279 PIER62298
Dyes, DyLight™
Protein Linking and Labeling Systems
DyLight™ Dyes have absorption spectra ranging from 350 nm to 770 nm and match the output wavelengths of common fluorescence instrumentation.
  • Available in both amine- and sulfhydryl-reactive chemistries for fast and efficient labeling of IgG or other proteins
  • High water solubility
  • Excellent photostability
  • Compatible with common fluorescence instrumentation
The DyLight™ dyes exhibit higher fluorescence intensity and photostability than Alexa Fluor®, CyDye® and LI-COR dyes in many applications and remain highly fluorescent over a broad pH range (pH 4 to 9). Additionally, the water solubility of the DyLight™ reagents allows a high dye-to-protein ratio without precipitation during conjugation.
Delivery information: Supplied as 1 mg or in convenience packs containing sufficient reagent for 5 single labelling reactions. Each single-use vial is sufficient to label 0,25 to 1 mg of protein (>45 kDa).
Order Now
SPECIFICATIONS
Environmentally Preferable
Host
Storage conditions - Store desiccated at –20 °C