Silver nitrate 0.1 N in aqueous solution USP Test Solution (TS)
Supplier: VWR Chemicals
Warning
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
85370.180EA 179 EUR
85370.180 85370.260
Silver nitrate 0.1 N in aqueous solution USP Test Solution (TS)
Silver nitrate
Ready to use solution.
  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 0
UN: 3264
ADR: 8 III
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM