Anti-IgA Goat Polyclonal Antibody (CF™ 488A)

Supplier: Biotium
20428-50UL 20428-500UL
BTIU20428-50ULEA 121 EUR
BTIU20428-50UL BTIU20428-500UL
Anti-IgA Goat Polyclonal Antibody (CF™ 488A)
Goat anti-human IgA reacts specifically with IgA heavy chains (α chains) and not with immunoglobulin light chains. To minimize crossreactivity, it has been adsorbed human IgG prior to conjugation. This conjugate is labeled with CF™488A dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Green fluorescent CF™488A (Ex/Em 490/515 nm) is a superior alternative to FITC (or fluorescein) owing to its exceptional brightness and photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR