Ethylmagnesium chloride 25% (w/w) 2.7 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
252578000 252571000
252578000.EA 263 EUR
252578000. 252571000.
Ethylmagnesium chloride 25% (w/w) 2.7 M in THF, AcroSeal®
Ethylmagnesium chloride
CAS No.: 2386-64-3
Formula: C₂H₅ClMg
MW: 88.82 g/mol
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000472
CAS Number: 2386-64-3
UN: 3399
ADR: 4.3 I
Order Now
Specification Test Results
Appearance Dark brown Solution
Titration with HCl 24 to 26 %