3-Chloroperoxybenzoic acid 70 - 75% 70-75% in water balance 3-Chlorobenzoic acid and water
Supplier: Thermo Fisher Scientific
Danger
255790010 255791000 255795000 255790250
255790010.EA 565 EUR
255790010. 255791000. 255795000. 255790250.
3-Chloroperoxybenzoic acid 70 - 75% 70-75% in water balance 3-Chlorobenzoic acid and water
3-Chloroperoxybenzoic acid
CAS No.: 937-14-4
Formula: C₇H₅ClO₃
Melting Pt: 92…94 °C
Density: 0.56 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00002127
CAS Number: 937-14-4
UN: 3106
ADR: 5.2,
Order Now
Specification Test Results
Appearance White Moist powder
Titration Iodometric 70 to 75 %
Water % balance (K.F.)
HPLC di-m-Chlorobenzoyl peroxide: < 0.1% Typical
3-Chlorobenzoic acid ≤12 %