N,N-Diethyl-p-phenylenediamine oxalate 96%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Bis(N,N-diethylbenzene-p-diamine) oxalate, DPD oxalate
407410100 407410250
ACRO407410100EA 111 EUR
ACRO407410100 ACRO407410250
N,N-Diethyl-p-phenylenediamine oxalate 96%
N,N-Diethyl-p-phenylenediamine oxalate
Formula: C₁₀H₁₆N₂·0.5 C₂H₂O₄
MW: 209.27 g/mol
Melting Pt: 145…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00070242
CAS Number: 62637-92-7
EINECS: 263-662-1
Order Now
Specification Test Results
Appearance Yellow to grey to light brown Crystalline powder
Infrared spectrum Conforms
Titration ≥95 % (DEPDA + oxalic acid)