N-Isopropylacrylamide 99% stabilised, pure

Supplier: Thermo Fisher Scientific
Warning

412780250 412781000 412785000 412780025
ACRO412780250EA 94.2 EUR
ACRO412780250 ACRO412781000 ACRO412785000 ACRO412780025
N-Isopropylacrylamide 99% stabilised, pure
N-Isopropylacrylamide

CAS No.: 2210-25-5

Formula: C₆H₁₁NO
Boiling Pt: 89…92 °C (2 mmHg)
Melting Pt: 62…66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041913
CAS Number: 2210-25-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥98.5 %
Stabilizer ca. 500 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR