Pivaloylacetonitrile 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4,4-Dimethyl-3-oxopentanenitrile, 4,4-Dimethyl-3-oxovaleronitrile, Trimethylacetylacetonitrile
418020250 418025000 418021000
ACRO418020250EA 34.6 EUR
ACRO418020250 ACRO418025000 ACRO418021000
Pivaloylacetonitrile 97%
Pivaloylacetonitrile
Appearance: White to cream Crystalline powder or crystals
Formula: C₇H₁₁NO
MW: 125.17 g/mol
Melting Pt: 66…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00010208
CAS Number: 59997-51-2
UN: 3439
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to cream Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 66 to 72 °C
Water ≤1 % (K.F.)
GC ≥96.0 %