2-(2-Pyridyl)benzimidazole 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: AI-4-88
167950050 167950100
ACRO167950050EA 64 EUR
ACRO167950050 ACRO167950100
2-(2-Pyridyl)benzimidazole 99%
2-(2-Pyridyl)benzimidazole
Appearance: Yellow to brown Powder
Formula: C₁₂H₉N₃
MW: 195.23 g/mol
Melting Pt: 220…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00005586
CAS Number: 1137-68-4
Order Now
Specification Test Results
Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 220 to 225 °C
HPLC ≥98.5 %