Janus green B, pure, high purity biological stain
Supplier: Thermo Fisher Scientific
191680050 191680250
ACRO191680050EA 62.3 EUR
ACRO191680050 ACRO191680250
Janus green B, pure, high purity biological stain
Janus green B
CAS No.: 2869-83-2
Formula: C₃₀H₃₁ClN₆
MW: 511.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011758
CAS Number: 2869-83-2
Order Now
Specification Test Results
Appearance Dark green to dark brown to black Powder
Infrared spectrum Conforms
Dye content ≥50 %