Glycyrrhizic acid monoammonium salt 98+%
Supplier: Thermo Fisher Scientific
Synonyms:
Monoammonium glycyrrhizinate, Glycyrrhizin monoammonium salt, Glycyrrhizin, ammonium salt
213750250
213751000
213750100
ACRO213750250EA
88.2
EUR
ACRO213750250
ACRO213751000
ACRO213750100
Glycyrrhizic acid monoammonium salt 98+%
Glycyrrhizic acid monoammonium salt
CAS No.: 53956-04-0
Formula:
C₄₂H₆₅NO₁₆ Melting Pt: 209 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00167400 CAS Number: 53956-04-0 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Water | ≤6.0 % (K.F.) |
Heavy metals | ≤20 ppm |
Sulfated ash | ≤0.2 % |
Related substances | ≤10.0 % 18-alpha-glycyrrhizic acid (HPLC) |
Specific optical rotation | +49.0° to +54.0° (20°C, 589 nm) (c=1, EtOH 50% v/v) (on anhydrous substance) |
Titration with HClO4 | 98.0 to 102.0 % (on anhydrous substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...