3-Ethoxymethacrolein 95% tech.
Supplier: Thermo Fisher Scientific
|
Warning
|
213990010
213990100
ACRO213990010EA
40.8
EUR
ACRO213990010
ACRO213990100
3-Ethoxymethacrolein 95% tech.
3-Ethoxymethacrolein
Appearance: Colorless to yellow Liquid
Formula:
C₆H₁₀O₂ Boiling Pt: 78…81 °C (14 mmHg) Density: 0.96 g/cm³ Flash Pt: 35 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006975 CAS Number: 42588-57-8 UN: 3271 ADR: 3,III |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4775 to 1.4815 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...