Aldosterone 98%
Supplier: Thermo Fisher Scientific
215360050 215360250 215361000
ACRO215360050EA 114 EUR
ACRO215360050 ACRO215360250 ACRO215361000
Aldosterone 98%
Aldosterone
CAS No.: 52-39-1
Formula: C₂₁H₂₈O₅
Melting Pt: 166.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00051136
CAS Number: 52-39-1
Order Now
Specification Test Results
Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +150° to +162° (20°C, 589 nm) (c=0.1, CHCl3)