Allyl methacrylate 98% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Methacrylic Acid Allyl Ester
215875000
215870025
ACRO215875000EA
68
EUR
ACRO215875000
ACRO215870025
Allyl methacrylate 98% stabilised
Allyl methacrylate
CAS No.: 96-05-9
Formula:
C₇H₁₀O₂ Boiling Pt: 144 °C (760 mmHg) Melting Pt: –65 °C Density: 0.93 g/cm³ Flash Pt: 37 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008592 CAS Number: 96-05-9 UN: 2929 ADR: 6.1,II |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4350 to 1.4370 (20°C, 589 nm) |
Water | ≤0.1 % |
GC | ≥97.5 % |
Color scale | ≤30 APHA |
Stabilizer | 200 to 250 ppm MEHQ |
Acids | ≤0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...