2-Iodoxybenzoic acid stabilised
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1-Hydroxy-1,2-benziodoxy-3-(1h)-one-1-xoide, IBX, 2-Iodylbenzoic acid
374650100 374650500
ACRO374650100EA 215 EUR
ACRO374650100 ACRO374650500
2-Iodoxybenzoic acid stabilised
2-Iodoxybenzoic acid
Appearance: White to off-white Powder or prills and/or agglomerates
Formula: C₇H₅IO₄
Melting Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD02912492
CAS Number: 61717-82-6
UN: 1759
Order Now
Specification Test Results
Appearance White to beige Powder or crystals
Titration Iodometric 39 to 50 % as 2-Iodoxybenzoic acid
Identification HPLC (2-Iodoxybenzoic acid, 2-Iodosobenzoic acid, Isophthalic acid, Benzoic acid)