n-Butylethylmagnesium 0.9 M in heptane, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
381181000 381188000
381181000.EA 108 EUR
381181000. 381188000.
n-Butylethylmagnesium 0.9 M in heptane, AcroSeal®
n-Butylethylmagnesium
CAS No.: 62202-86-2
Formula: C₆H₁₄Mg
Density: 0.73 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00072569
CAS Number: 62202-86-2
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Clear to slightly hazy colorless Liquid
Titration 13.5 to 14.5 % (on Mg basis)