2-Amino-5-methylthiazole 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 5-Methylthiazol-2-ylamine
392790050 392790250 392791000
ACRO392790050EA 23.4 EUR
ACRO392790050 ACRO392790250 ACRO392791000
2-Amino-5-methylthiazole 98%
2-Amino-5-methylthiazole
Formula: C₄H₆N₂S
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00078317
CAS Number: 7305-71-7
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Brown Powder
Infrared spectrum Conforms
GC ≥97.5 %