Zinc sulphate monohydrate 99%
Supplier: Thermo Fisher Scientific
Danger
389800010 389800050 389802500
ACRO389800010EA 88.1 EUR
ACRO389800010 ACRO389800050 ACRO389802500
Zinc sulphate monohydrate 99%
Zinc sulphate monohydrate
Formula: ZnSO₄·1H₂O
MW: 179.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149893
CAS Number: 7446-19-7
UN: 3077
ADR: 9 III
Merck Index: 13,10213
Order Now
Specification Test Results
Appearance White Powder
Titration Complexometric ≥98.5 %