Dibutylamine for HPLC
Supplier: Thermo Fisher Scientific
Danger
390610100 390611000
ACRO390610100EA 42.6 EUR
ACRO390610100 ACRO390611000
Dibutylamine for HPLC
Dibutylamine
Formula: C₈H₁₉N
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –62 °C
Density: 0.76 g/cm³
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00009429
CAS Number: 111-92-2
UN: 2248
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d7d86589224f8565e12dc8e6b30a8de923393f&vid=d2cc023db25785e54555fc105aa8436e65fd9cbc&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Water ≤0.1 %
GC ≥99.0 %
Residue after evaporation ≤0.0005 %
UV transmittance ≥99 % (c=0.01M water, 254 nm)