2-Iodopyridine 98%
Supplier: Thermo Fisher Scientific
Warning
379330010 379330050
ACRO379330010EA 37.8 EUR
ACRO379330010 ACRO379330050
2-Iodopyridine 98%
2-Iodopyridine
Formula: C₅H₄IN
MW: 205 g/mol
Boiling Pt: 52 °C (.9 mmHg)
Melting Pt: 118…120 °C
Density: 1.928 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00464928
CAS Number: 5029-67-4
Order Now
Specification Test Results
Appearance Clear colorless to golden Liquid
Infrared spectrum Conforms
GC ≥97.5 %