Peqlab, Electroporation Cuvettes
Supplier: VWR Peqlab
732-2924EA 97.2 EUR
732-2924 732-2922 732-2920 732-2918 732-2916
Peqlab, Electroporation Cuvettes
Cuvettes Electroporation Cuvettes
High precision manufacturing and strict quality control guarantee consistent constant electroporation.
  • Coloured caps for selecting the correct column widths
  • 4 and 2 mm cuvettes for long electrodes
  • Sterilised with gamma rays
Packaging: Individually packed to ensure sterility.