Safety helmets, uvex pheos IES
Supplier: UVEX
9772.040 9772.140 9772.240 9772.340
111-1878EA 23.5 EUR
111-1878 111-1907 111-1908 UVEX9772.340
Safety helmets, uvex pheos IES
Head Protection Safety Helmets
This helmet features IES, an Integrated Eyewear System, which enables users to attach pheos safety spectacles to the helmet. The spectacles can be pushed up inside the uvex pheos helmet or pulled down to sit on the nose. The sliding element is jointed so that movement of the helmet does not impact the eyewear.
  • Three variable air vents aid airflow and help regulate the temperature within the helmet, making it more comfortable to wear
  • Adapter systems for attaching uvex visors, uvex earmuffs and lamps
  • Interior harness makes it possible to attach the uvex pheos spectacles using the spectacle mounting clip
  • Helmet can be individually adjusted using the rotating ratchet wheel to alter the width size
  • Suitable for ambient temperatures down to −30 °C and for molten metal (splash)
Size: 550 to 610 mm
EN 397, low temperature (–30 °C), and molten metal
Certifications: EN 397, low temperature (–30 °C), and molten metal.
Accessories information: Highly reflective sticker set for all uvex pheos and uvex pheos alpine helmets. Prism foil ensures superior luminosity in low light environments.