ESD occupational shoes, slip-on, Zwiesel
Supplier: SCHURR SCHUHVERTRIEB
1.1118.0151/41 1.1018.0151/36 1.1018.0151/37 1.1018.0151/38 1.1018.0151/39 1.1018.0151/40 1.1118.0151/42 1.1118.0151/43 1.1118.0151/44 1.1118.0151/45 1.1118.0151/46
114-7170EA 77.8 EUR
114-7170 114-7165 114-7166 114-7167 114-7168 114-7169 114-7171 114-7172 114-7173 114-7174 114-7175
ESD occupational shoes, slip-on, Zwiesel
Shoes and Boots
Comfortable ESD slip on shoes with microfibre upper and a PU sole.
  • DRYtech® textile lining ensures excellent foot climate
  • Insole with antibacterial treatment reduces odour and bacterial growth
  • ESD compliant, slip-resistant sole
Suitable for use in kitchens and food-processing industry.
Colour: White.
Certifications: EN ISO 20347:2012; O2, SRA