Cut resistant gloves, uvex C500 XG
Supplier: UVEX
6060007 6060008 6060009 6060010 6060011
112-4570EA 30.8 EUR
112-4570 112-4571 112-4572 112-4573 112-4574
Cut resistant gloves, uvex C500 XG
Gloves
These bamboo-rayon/Dyneema®/glass/polyamide gloves feature a full HPE Xtra Grip coating. The Bamboo TwinFlex® technology ensures maximum cut resistance, exceptional abrasion resistance and outstanding wearing comfort.
  • Xtra Grip coating provides a secure grip in oily and wet environments
  • Palm, fingers and back of the hand are fully coated
  • Outstanding wearing comfort and tactile feel with climazone technology
  • Oil and grease resistant
  • Highest cut resistance level 5
  • Silicone-free according to imprint test
Typical applications include: Automotive industry, construction, brewery/beverage industry, glass industry, maintenance/servicing, metal industry, assembly works, paper industry, shipping/logistics, transportation works.
Certifications: EN 388:2016 (4X42C)
OEKO-Tex® Standard 100