Calcium chloride 96%, granules for analysis
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
219170025 219171000 219175000
ACRO219170025EA 496 EUR
ACRO219170025 ACRO219171000 ACRO219175000
Calcium chloride 96%, granules for analysis
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: >1600 °C (760 mmHg)
Melting Pt: 782 °C
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
Merck Index: 14,01659
Order Now
Specification Test Results
Appearance White Granules
Titration Complexometric ≥95 %
Fluoride (F) ≤40 ppm
Sulfate (SO4) ≤0.2 %
Arsenic (As) ≤1 ppm
Barium (Ba) ≤0.4 %
Iron (Fe) ≤50 ppm
Lead (Pb) ≤5 ppm
Mercury (Hg) ≤1 ppm