tert-Butylmagnesium chloride 1.7 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
331671000 331678000
331671000.EA 89.7 EUR
331671000. 331678000.
tert-Butylmagnesium chloride 1.7 M in THF, AcroSeal®
tert-Butylmagnesium chloride
Formula: C₄H₉ClMg
MW: 116.87 g/mol
Density: 0.931 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000465
CAS Number: 677-22-5
UN: 3399
ADR: 4.3 I
Order Now
Specification Test Results
Appearance Dark brown Solution
Titration Active base 19 to 21 %