9-Vinylcarbazole 97%
Supplier: Thermo Fisher Scientific
Warning
387220050 387220250 387221000
ACRO387220050EA 30.2 EUR
ACRO387220050 ACRO387220250 ACRO387221000
9-Vinylcarbazole 97%
9-Vinylcarbazole
Formula: C₁₄H₁₁N
Boiling Pt: 154 °C (3 mmHg)
Melting Pt: 63…65 °C
Flash Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00004966
CAS Number: 1484-13-5
UN: 2811
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Off-white to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥96.0 %