Sodium mercury amalgam Na ∼5%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Mercury sodium alloy
388590100 388590500
ACRO388590100EA 173 EUR
ACRO388590100 ACRO388590500
Sodium mercury amalgam Na ∼5%
Sodium mercury amalgam (Mercury Sodium alloy (80:20 w%))
Formula: Hg:Na
Storage Temperature: Ambient
MDL Number: MFCD00210362
CAS Number: 11110-52-4
UN: 3401
ADR: 4.3 I
Order Now
Specification Test Results
Appearance Silver to grey Irregular pieces
Sodium (Na) 5 % (approximately)