Ammonium trifluoromethanesulphonate 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Methanesulphonic acid,1,1,1-trifluoro-, ammonium salt (1:1), Ammonium triflate
388730050 388730250 388730500
ACRO388730050EA 67.8 EUR
ACRO388730050 ACRO388730250 ACRO388730500
Ammonium trifluoromethanesulphonate 99%
Ammonium trifluoromethanesulphonate
Formula: CH₄F₃NO₃S
MW: 167.102 g/mol
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00075332
CAS Number: 38542-94-8
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥98.5 %