Calcium acetylide 72-82
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Calcium acetylide
389792500 389790010 389790025
ACRO389792500EA 34.6 EUR
ACRO389792500 ACRO389790010 ACRO389790025
Calcium acetylide 72-82
Calcium acetylide
Formula: CaC₂
MW: 64.1 g/mol
Boiling Pt: 2300 °C (760 mmHg)
Melting Pt: 2160 °C
Density: 2.22
Storage Temperature: Ambient
MDL Number: MFCD00010905
CAS Number: 75-20-7
UN: 1402
ADR: 4.3 I
Merck Index: 12,01696
Order Now
Specification Test Results
Appearance Dark grey Lumps
Titration Complexometric 12 to 23 % (CaO)
Particle size 7 to 25 mm