O,O'-Di-2-pyridyl thiocarbonate 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Di-2-Pyridyl Thionocarbonate, Thiocarbonic Acid O,O'-Di-2-pyridyl Ester
390880010 390880050
ACRO390880010EA 69.6 EUR
ACRO390880010 ACRO390880050
O,O'-Di-2-pyridyl thiocarbonate 97%
O,O'-Di-2-pyridyl thiocarbonate
Formula: C₁₁H₈N₂O₂S
MW: 232.26 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Freezer
MDL Number: MFCD00074870
CAS Number: 96989-50-3
Order Now
Specification Test Results
Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %