Diammonium thiosulphate 96%, pure
Supplier: Thermo Fisher Scientific
Synonyms: Ammonium hyposulphite, Ammonium thiosulphate
391660025 391660100 391665000
ACRO391660025EA 109 EUR
ACRO391660025 ACRO391660100 ACRO391665000
Diammonium thiosulphate 96%, pure
di-Ammonium thiosulphate
Formula: H₈N₂O₃S₂
MW: 148.21 g/mol
Melting Pt: 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00015965
CAS Number: 7783-18-8
Merck Index: 13,00565
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals and/or chunks
Titration Iodimetric ≥95 %