n-Octane, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
396900010 396901000
396900010.EA 484 EUR
396900010. 396901000.
n-Octane, extra dry 99+%, AcroSeal®
n-Octane
Formula: H₃C(CH₂)₆CH₃
Boiling Pt: 125…127 °C (760 mmHg)
Melting Pt: –57 °C
Density: 0.708 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009556
CAS Number: 111-65-9
UN: 1262
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68adf890d68728825d4a2fa4b032700b93f710b8&vid=b00a2925902bc425c80f8ebb3f0e7b456c800ae3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.002 %