3-Amino-4-methylpyridine 97%
Supplier: Thermo Fisher Scientific
Danger
429860050 429860250
ACRO429860050EA 41.7 EUR
ACRO429860050 ACRO429860250
3-Amino-4-methylpyridine 97%
3-Amino-4-methylpyridine
Formula: C₆H₈N₂
Melting Pt: 105…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00128871
CAS Number: 3430-27-1
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 100 to 108 °C
HPLC ≥96.0 %
Uniformity coefficient ≤2.00
Moisture holding capacity 60 to 70 % (H)
Total volume capacity ≥2.60 eq/l (H)