1H-Indazole-6-carboxaldehyde 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1H-Indazole-6-carbaldehyde, 6-Formylindazole, INDAZOLE-6-CARBOXALDEHYDE, 6-Formyl-1H-indazole

431040050 431040010
ACRO431040050EA 231 EUR
ACRO431040050 ACRO431040010
1H-Indazole-6-carboxaldehyde 97%
1H-Indazole-6-carboxaldehyde
Formula: C₈H₆N₂O
Melting Pt: 180.6…180.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06738282
CAS Number: 669050-69-5

Order Now

Specification Test Results

Appearance White to yellow to brown Powder
Infrared spectrum Conforms
Melting point 180 to 185 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR