Trioctylsilane 95%
Supplier: Thermo Fisher Scientific
Warning
434900050 434900250
ACRO434900050EA 57.4 EUR
ACRO434900050 ACRO434900250
Trioctylsilane 95%
Trioctylsilane
Formula: C₂₄H₅₂Si
MW: 368.76 g/mol
Boiling Pt: 163…165 °C (.2 mmHg)
Density: 0.821 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00053812
CAS Number: 18765-09-8
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %