Chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Chlorodihydrido[bis(2-di-i-propylphosphinoethyl)amine]iridium(III), CHLORODIHYDRIDO[BIS(2-DI-I-PROPYLPHOSPHINOETHYL)AMINE]IRIDIUM (III), CHLORODIHYDRIDO[BIS(2-DI-I-PROPYLPHOSPHINOETHYL)AMINE]IRIDIUM(III)
435120010 435122500
ACRO435120010EA 299 EUR
ACRO435120010 ACRO435122500
Chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium 97%
Chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium
Formula: C₁₆H₄₂ClIrN₂P₂
MW: 552.141 g/mol
Melting Pt: 240.6…241.9 °C
Storage Temperature: Ambient
MDL Number: MFCD07782018
CAS Number: 791629-96-4
Order Now
Specification Test Results
Appearance White to tan Powder
Infrared spectrum Conforms
Elemental Analysis 2.50 to 2.70 % (N)